missing translation for 'onlineSavingsMsg'
Learn More

CXCL2/GRO beta/MIP-2/CINC-3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18340242 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340242 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340242 Supplier Novus Biologicals Supplier No. NBP299401AF350

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL2/GRO beta/MIP-2/CINC-3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CXCL2/GRO beta/MIP-2/CINC-3 (NP_002080.1; Ala35-Asn107).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2920
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.