missing translation for 'onlineSavingsMsg'
Learn More

CXCL15/Lungkine Antibody (RM0207-16M7), DyLight 594, Novus Biologicals™

Product Code. 18412426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412426 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412426 Supplier Novus Biologicals Supplier No. NBP212416DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCL15/Lungkine Monoclonal antibody specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL15/Lungkine
Applications Western Blot
Classification Monoclonal
Clone RM0207-16M7
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Il8, IL-8, lungkine, Scyb15, weche
Host Species Rat
Immunogen Mouse recombinant CXCL15/Lungkine
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20309
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.