missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL15/Lungkine Antibody (RM0207-16M7), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-12416CL1
This item is not returnable.
View return policy
Description
CXCL15/Lungkine Monoclonal antibody specifically detects CXCL15/Lungkine in Mouse samples. It is validated for Western Blot
Specifications
| CXCL15/Lungkine | |
| Monoclonal | |
| CoraFluor 1 | |
| Il8, IL-8, lungkine, Scyb15, weche | |
| Mouse recombinant CXCL15/Lungkine | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0207-16M7 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 20309 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction