Learn More
CXCL14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10468-1-AP-150UL
 View more versions of this product
Product Code. 16876863
  • £349.00 / 150µL
Description
This gene belongs to the cytokine gene family which encode secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC (Cys-X-Cys) subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes but not for lymphocytes, dendritic cells, neutrophils or macrophages. It has been implicated that this cytokine is involved in the homeostasis of monocyte-derived macrophages rather than in inflammation.
Specifications
Specifications
CXCL14
Polyclonal
Unconjugated
CXCL14
BMAC, bolekine, BRAK, C X C motif chemokine 14, Chemokine BRAK, CXCL14, Kec, KS1, MIP 2g, MIP2G, NJAC, PSEC0212, SCYB14, Small inducible cytokine B14
Rabbit
Antigen Affinity Chromatography
RUO
9547
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95715
CXCL14
CXCL14 Fusion Protein Ag0747
150 μL