missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CXCL13 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P141BT-50UG

 View more versions of this product

Product Code. 17844783

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: VLEVYYTSLR CRCVQESSVF IPRRFIDRIQ ILPRGNGCPR KEIIVWKKNK SIVCVDPQAE WIQRMMEVLR KRSSSTLPVP VFKRKIP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BCA-1 (CXCL13). Anti-Human BCA-1 (CXCL13)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human BCA-1 (CXCL13) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BCA-1 (CXCL13) (500-P141) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BCA-1 (CXCL13). Western Blot: To detect hBCA-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBCA-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P141BT-1MG will be provided as 2 x 500 μg

CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt's Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL13
Polyclonal
Biotin
Cxcl13
4631412M08Rik; Angie; ANGIE2; B cell-attracting chemokine 1; b lymphocyte chemoattractant; BCA1; BCA-1; B-cell chemoattractant; B-cell-attracting chemokine 1; B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1); BLC; BLR1L; B-lymphocyte chemoattractant; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chem; chemokine (C-X-C motif) ligand 13; chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant); CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; C-X-C motif chemokine 13; C-X-C motif chemokine ligand 13; CXCL; Cxcl13; SCYB13; small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cell chemoattractant); small inducible cytokine subfamily B (Cys-X-Cys), member 13; small-inducible cytokine B13
Rabbit
Antigen affinity chromatography
RUO
10563
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
O43927
Cxcl13
E.coli-derived Recombinant Human BCA-1 (CXCL13).
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.