missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL13 Monoclonal Antibody (53610), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523666
This item is not returnable.
View return policy
Description
In ELISAs, this antibody does not cross-react with recombit human (rh) CXCL1, rhCXCL2, rhCXCL3, rhCXCL8/IL-8, rhCXCL10, rhCXCL9, rhCXCL12/SDF-1 alpha, or rhCXCL12/SDF-1 beta.
B lymphocyte chemoattractant, independently cloned and named Angie, is a CXC chemokine strongly expressed in the follicles of the spleen, lymph nodes, and Peyer′s patches. It preferentially promotes the migration of B lymphocytes (compared to T cells and macrophages), apparently by stimulating calcium influx into, and chemotaxis of, cells expressing Burkitt′s lymphoma receptor 1 (BLR-1). It may therefore function in the homing of B lymphocytes to follicles.
Specifications
| CXCL13 | |
| Monoclonal | |
| PE | |
| CXCL13 | |
| ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13, B-cell chemoattractant, B-cell-attracting chemokine 1, B-cell-homing chemokine (ligand for Burkitt′s lymphoma receptor-1), B-lymphocyte chemoattractant, C-X-C motif chemokine 13, CXC chemokine BLC, b cell-attracting chemokine 1, b lymphocyte chemoattractant, chem | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10563 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 53610 | |
| Buffered saline solution with BSA and 0.09% sodium azide | |
| O43927 | |
| CXCL13 | |
| E. coli-derived recombit human CXCL13/BLC/BCA-1 Val23-Arg94 | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction