missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17402-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can detect all the isoforms of CXCL12/SDF1.
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| CXCL12/SDF-1 | |
| Polyclonal | |
| Unconjugated | |
| CXCL12 | |
| C X C motif chemokine 12, CXCL12, hIRH, hSDF 1, IRH, PBSF, SCYB12, SDF 1, SDF 1a, SDF 1b, SDF1, SDF1A, SDF1B, Stromal cell derived factor 1, TLSF a, TLSF b, TPAR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6387 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P48061 | |
| CXCL12 | |
| CXCL12/SDF-1 Fusion Protein Ag11379 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction