missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 beta Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-05880CL1
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 beta Polyclonal antibody specifically detects CXCL12/SDF-1 beta in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CXCL12/SDF-1 beta | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| hSDF-1beta, SDF 1beta, SDF1B | |
| Produced in rabbits immunized with purified, recombinant Human CXCL12/SDF-1 beta (Accession#: P48061-1; Lys22-Met93) | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 6387 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction