missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCL12/SDF-1 beta Polyclonal specifically detects CXCL12/SDF-1 beta in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | CXCL12/SDF-1 beta |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hSDF-1beta, SDF 1beta, SDF1B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Mouse CXCL12/SDF-1 beta (P40224-1; Lys22-Lys89). |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?