missing translation for 'onlineSavingsMsg'
Learn More

CXCL12/SDF-1 Antibody (79014) [PE], Novus Biologicals™

Product Code. 30736417 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736417 Supplier Novus Biologicals Supplier No. FAB310P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human,Mouse samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL12/SDF-1
Applications Neutralization
Classification Monoclonal
Clone 79014
Conjugate PE
Formulation PBS
Gene Alias chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL12/SDF-1 alpha , Lys22-Lys89, Accession # Q6ICW0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.