missing translation for 'onlineSavingsMsg'
Learn More

CXCL12/SDF-1 alpha Antibody, DyLight 550, Novus Biologicals™

Product Code. 18411838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411838 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411838 Supplier Novus Biologicals Supplier No. NBP229480R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CXCL12/SDF-1 alpha Polyclonal antibody specifically detects CXCL12/SDF-1 alpha in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL12/SDF-1 alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CXCL12 chemokine (C-X-C motif) ligand 12, IRH, PBSF, SCYB12, SDF1, SDF1A, SDF1B, TLSF, TPAR1
Host Species Rabbit
Immunogen Antisera to human SDF 1 alpha were raised by repeated immunisations of rabbits with highly purified antigen. Purified IgG prepared by affinity chromatography.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6387
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.