missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CXCL12 Monoclonal Antibody (K92033_5D11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556256
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CXCL12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P48061 | |
| CXCL12 | |
| Recombinant human CXCL12 (immunogen range is 22-93). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K92033_5D11 | |
| Unconjugated | |
| CXCL12 | |
| 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; hIRH; hSDF-1; Intercrine reduced in hepatomas; IRH; PBSF; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; Scyb12; Sdf1; SDF-1; SDF-1 gamma; SDF1A; SDF-1a; SDF-1-alpha(3-67); SDF1B; SDF-1b; SDF-1-beta(3-72); Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tlsfa; TLSF-a; Tlsfb; TLSF-b; Tpar1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6387 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction