missing translation for 'onlineSavingsMsg'
Learn More

CXCL12 alpha (SDF-1 alpha) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen 500-P164GBT-1MG

 View more versions of this product

Product Code. 17834783

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK. Preparation: Produced from sera of goats immunized with highly pure recombinant Murine SDF-1alpha (CXCL12). Anti-Murine SDF-1alpha (CXCL12)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine SDF-1alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine SDF-1alpha (500-P164G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine SDF-1alpha. Western Blot: To detect Murine SDF-1alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Murine SDF-1alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P164GBT-1MG will be provided as 2 x 500 μg

CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL12 alpha (SDF-1 alpha)
Polyclonal
Biotin
CXCL12
12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 a; SDF 1 alpha; SDF 1a; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1
Goat
Antigen affinity chromatography
RUO
20315
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P40224
CXCL12
E.coli-derived, 7.9 kDa Recombinant Murine SDF-1alpha (CXCL12)
2 x 500 μg
Primary
Mouse
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.