Learn More
CXCL11 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10707-1-AP-150UL
This item is not returnable.
View return policy
Description
The CXCL11 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL11. This antibody recognizes human antigen. The CXCL11 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| CXCL11 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6373 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human |