missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL11/I-TAC Mouse anti-Human, Clone: B-S40, Novus Biologicals™
Mouse Monoclonal Antibody
£305.00 - £571.00
Specifications
| Antigen | CXCL11/I-TAC |
|---|---|
| Clone | B-S40 |
| Concentration | 1 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369923
|
Novus Biologicals
NBP3-18117-200UL |
200 μg |
£305.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399396
|
Novus Biologicals
NBP3-18117-500UL |
500 μg |
£571.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Human samples. It is validated for ELISASpecifications
| CXCL11/I-TAC | |
| 1 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| PBS. Sterile-filtered through 0.22 um. | |
| 6373 | |
| IgG1 κ | |
| Affinity purified |
| B-S40 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
| Recombinant human CXCL11/I-TAC. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title