Learn More
CXCL10/IP10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10937-1-AP-150UL
 View more versions of this product
Product Code. 16897653
  • £349.00 / 150µL
Description
The CXCL10/IP10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL10/IP10. This antibody recognizes human antigen. The CXCL10/IP10 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
CXCL10/IP10
Polyclonal
Unconjugated
150 μL
3627
IgG
Western Blot, Immunohistochemistry
1053 μg/mL