missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IP-10/CXCL10 Mouse Matched Antibody Pair

Product Code. 15591977
Change view
Click to view available options
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15591977 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15591977 Supplier Invitrogen™ Supplier No. BMS6018MST

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1 vial (0.275mL) polyclonal Coating Antibody to mouse IP-10 (100μg/mL)
  • 1 vial (55μL) Biotin-Conjugate anti-mouse IP-10 polyclonal antibody
  • 1 vial (22μL) Streptavidin-HRP
  • 10 vials mouse IP-10 Standard protein lyophilized, 1ng/mL upon reconstitution
  • 2 vials (50mL) Sample Diluent

Matched Antibody Pair

The Mouse IP-10 Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,sample dilutent and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IP-10. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

CXCL10 ( C-X-C Motif Chemokine Ligand 10) is a 10 kDa protein and member of the CXC family that acts by binding to CXCR3. Binding of CXCL10 to CXCR3 results in pleiotropic effects,including stimulation of monocytes,natural killer and T-cell migration,and modulation of adhesion molecule expression. CXCL10 is a pro-inflammatory chemoattractant cytokine that attracts activated T lymphocytes. CXCL10 is constitutively expressed at low levels in lymph organs,and the expression is highly induced by IFN-gamma and to some extent by IL12. It is an important biomarker for diagnosis of many clinical conditions,and it plays an important role in host defense and inflammation. CXCL10 exhibits an angiostatic and an antitumor role that is also involved in Th1 inflammatory diseases,Hashimoto′s thyroiditis,Graves′ disease and Type 1 diabetes mellitus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL10 (IP-10)
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Gene Accession No. P17515
Gene Alias Interferon-gamma-inducible protein 10, CXCL10
Gene Symbols Cxcl10
Quantity 10 x 96 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15945
Target Species Mouse
Content And Storage -20°C
Product Type ELISA Kit
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.