Learn More
Invitrogen™ CXCL1 (GRO alpha) Monoclonal Antibody (KTYFLF), eFluor™ 660, eBioscience™
Mouse Monoclonal Antibody
£258.00
Specifications
| Antigen | CXCL1 (GRO alpha) |
|---|---|
| Clone | KTYFLF |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: This KTYFLF monoclonal antibody reacts with human CXCL1 and has no cross-reactivity to human CXCL2 or CXCL3. CXCL1, or GRO (growth-related oncogene) alpha, is a pro-inflammatory CXC chemokine first identified by its constitutive overexpression in some tumors. It is closely related to CXCL2 (GRO beta) and CXCL3 (GRO gamma), with which it shares 90% and 86% sequence homology, respectively. These proteins, along with IL-8 or CXCL8, were later found to be critical for neutrophil mobilization and degranulation, as well as vascular permeabilization and angiogenesis. Signaling occurs through the G protein-coupled receptor CXCR2, which is shared with GRO beta and gamma. CXCR2 can also be activated by IL-8, although some studies suggest that the majority of IL-8 activity on neutrophils is mediated by CXCR1. CXCL1 is secreted by monocytes, epithelial cells, and fibroblasts in response to pro-inflammatory stimuli such as LPS, IL-1 beta, and TNF alpha. Overexpression is observed in many malignant tumors, where it contributes to tumor vascularization and metastasis. Applications Reported: This KTYFLF antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This KTYFLF antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of peripheral blood monocytes. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that wi...
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.Specifications
| CXCL1 (GRO alpha) | |
| 5 μL/Test | |
| Flow Cytometry | |
| eFluor 660 | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P09341 | |
| 2919 | |
| Primary | |
| Affinity chromatography |
| KTYFLF | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Cxcl1 | |
| alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51 | |
| Cxcl1 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.