missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL1/GRO alpha/KC/CINC-1 Antibody (20326) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB275JF585
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human,Primate samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
| E. coli-derived recombinant human CXCL1/GRO alpha /KC/CINC-1 | |
| 0.1 mL | |
| Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Membrane Trafficking and Chaperones, Signal Transduction | |
| 2919 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), CyTOF | |
| 20326 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction