missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX516, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
CX516 (BDP 12) is an ampakine and acts as an AMPA receptor positive allosteric modulator for the research of Alzheimer's disease, schizophrenia and mild cognitive impairment (MCI).
Specifications
Specifications
| Chemical Name or Material | CX516 |
| Physical Form | Solid |
| CAS | 154235-83-3 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H15N3O |
| For Use With (Application) | Neuroscience-Neurodegeneration |
| Synonym | BDP 12 |
| Solubility Information | H2O : 100 mg/mL (414.44 mM; Need ultrasonic) ∣DMSO : ≥ 41 mg/mL (169.92 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction