missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CR1 Rat anti-Human , Alexa Fluor 647, Clone: 2A9-1, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 565895
This item is not returnable.
View return policy
Description
CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow Cytometry
Specifications
| CX3CR1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor | |
| Human CX3CR1 Recombinant Protein | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| 2A9-1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction