missing translation for 'onlineSavingsMsg'
Learn More

CX3CR1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18410825 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410825 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410825 Supplier Novus Biologicals Supplier No. NBP176872JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CX3CR1 Polyclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CX3CR1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKBRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28C-X3-C CKR-1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to amino acids in a extracellular loop of human CX3CR1. The immunogen is located within amino acids 160 - 210 of CX3CR1. Amino Acid Squence: CLGDYPEVLQEIWP
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, GPCR, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 1524
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.