missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | CX3CR1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CX3CR1 | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Chemokines and Cytokines, GPCR | |
| PBS with 0.02% Sodium Azide | |
| Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKBRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28C-X3-C CKR-1 | |
| CX3CR1 | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of CX3CR1 are known to exist. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P49238 | |
| 1524 | |
| Antibody was raised against a peptide corresponding to amino acids in a extracellular loop of human CX3CR1. The immunogen is located within amino acids 160 - 210 of CX3CR1. Amino Acid Squence: CLGDYPEVLQEIWP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title