missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CX3CL1/Fractalkine Antibody (quetmolimab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28244-50ug
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Monoclonal antibody specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CX3CL1/Fractalkine | |
| Monoclonal | |
| Unconjugated | |
| P78423 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| quetmolimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| CX3CL1/ Fractalkine | |
| 50 μg | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 6376 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction