missing translation for 'onlineSavingsMsg'
Learn More

CX-6258 hydrochloride hydrate, MedChemExpress

Product Code. 16497458
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497458 10 mg 10mg
16443487 50 mg 50mg
16419947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497458 Supplier Medchem Express Supplier No. HY18095A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CX-6258 hydrochloride hydrate is a potent and kinase selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM and 16 nM for Pim-1, Pim-2 and Pim-3, respectively.

CX-6258 hydrochloride hydrate is a potent and kinase selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM and 16 nM for Pim-1, Pim-2 and Pim-3, respectively.

Specifications

Chemical Name or Material CX-6258 hydrochloride hydrate
CAS 1353858-99-7
Color Orange
Purity Grade Notes Research
Molecular Formula C26H27Cl2N3O4
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (48.41 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (13.83 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1NC2=CC=C(Cl)C=C2/C1=C\C3=CC=C(C4=CC=CC(C(N5CCCN(C)CC5)=O)=C4)O3.[H]Cl.O
Molecular Weight (g/mol) 516.42
Formula Weight 516.42
Percent Purity 98.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.