missing translation for 'onlineSavingsMsg'
Learn More

CX-5461, MedChemExpress

Product Code. p-200096215
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488337

Brand: Medchem Express HY13323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CX-5461 is a potent and oral rRNA synthesis inhibitor. It inhibits RNA polymerase I-driven transcription of rRNA with IC50s of 142, 113, and 54 nM in HCT-116, A375, and MIA PaCa-2 cells, respectively.

CX-5461 is a potent and oral rRNA synthesis inhibitor. It inhibits RNA polymerase I-driven transcription of rRNA with IC50s of 142, 113, and 54 nM in HCT-116, A375, and MIA PaCa-2 cells, respectively.

Specifications

Chemical Name or Material CX-5461
CAS 1138549-36-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H27N7O2S
Quantity 5 mg
Solubility Information DMSO : 2 mg/mL (3.89 mM; Need ultrasonic)
SMILES CC1=NC=C(CNC(C2=C3N(C4=NC(N5CCN(CCC5)C)=CC=C4C2=O)C6=C(S3)C=CC=C6)=O)N=C1
Molecular Weight (g/mol) 513.61
Formula Weight 513.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.