missing translation for 'onlineSavingsMsg'
Learn More

CX-5461, MedChemExpress

Product Code. 16488337
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488337 5 mg 5mg
16425648 10 mg 10mg
16411817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488337 Supplier Medchem Express Supplier No. HY13323
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CX-5461 is a potent and oral rRNA synthesis inhibitor. It inhibits RNA polymerase I-driven transcription of rRNA with IC50s of 142, 113, and 54 nM in HCT-116, A375, and MIA PaCa-2 cells, respectively.

CX-5461 is a potent and oral rRNA synthesis inhibitor. It inhibits RNA polymerase I-driven transcription of rRNA with IC50s of 142, 113, and 54 nM in HCT-116, A375, and MIA PaCa-2 cells, respectively.

Specifications

Chemical Name or Material CX-5461
CAS 1138549-36-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H27N7O2S
Quantity 5 mg
Solubility Information DMSO : 2 mg/mL (3.89 mM; Need ultrasonic)
SMILES CC1=NC=C(CNC(C2=C3N(C4=NC(N5CCN(CCC5)C)=CC=C4C2=O)C6=C(S3)C=CC=C6)=O)N=C1
Molecular Weight (g/mol) 513.61
Formula Weight 513.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.