missing translation for 'onlineSavingsMsg'
Learn More

CX 546, TRC

Product Code. 30357470
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357470 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357470 Supplier Toronto Research Chemicals Supplier No. TRCC85800010MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material CX 546
CAS 215923-54-9
Molecular Formula C14 H17 N O3
Quantity 10 mg
InChI Formula InChI=1S/C14H17NO3/c16-14(15-6-2-1-3-7-15)11-4-5-12-13(10-11)18-9-8-17-12/h4-5,10H,1-3,6-9H2
SMILES O=C(N1CCCCC1)c2ccc3OCCOc3c2
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-yl(piperidin-1-yl)methanone
Molecular Weight (g/mol) 247.29
Formula Weight 247.1208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.