missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CX 546
GREENER_CHOICE

Product Code. 18074712
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074712

Brand: Tocris Bioscience™ 2980/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AMPA receptor potentiator

AMPA receptor potentiator. Binds specifically to the agonist bound non-desensitized receptor, most likely destabilizing the desensitized receptor conformation. Enhances cognitive function in rats. Neuroprotective in mice undergoing repeat ketamine anesthesia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215923-54-9
Molecular Formula C14H17NO3

Specifications

Quantity 50 mg
Formula Weight 247.29
Purity >99%
Molecular Weight (g/mol) 247.294
Chemical Name or Material CX 546
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.