missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CWC22 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92106-0.1ml
This item is not returnable.
View return policy
Description
CWC22 Polyclonal antibody specifically detects CWC22 in Mouse samples. It is validated for Western Blot
Specifications
| CWC22 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| CWC22 spliceosome-associated protein homolog (S. cerevisiae), EIF4GL, fSAPbpre-mRNA-splicing factor CWC22 homolog, KIAA1604NCMfunctional spliceosome-associated protein b, Nucampholin homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 639-908 of human CWC22 (NP_065994.1). LTDELREHLKNTPKVIVAQKPDVEQNKSSPSSSSSASSSSESDSSDSDSDSSDSSSESSSEESDSSSISSHSSASANDVRKKGHGKTRSKEVDKLIRNQQTNDRKQKERRQEHGHQETRTERERRSEKHRDQNSSGSNWRDPITKYTSDKDVPSERNNYSRVANDRDQEMHIDLENKHGDPKKKRGERRNSFSENEKHTHRIKDSENFRRKDRSKSKEMNRKHSGSRSDEDRYQNGAERRWEKSSRYSEQSRESKKNQDRRREKSPAKQK | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57703 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction