missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CWC15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25293-1-AP-150UL
This item is not returnable.
View return policy
Description
Cwc15 gene ontology annotations related to this gene include mRNA splicing, via spliceosome.Specifications
| CWC15 | |
| Polyclonal | |
| Unconjugated | |
| CWC15 | |
| C11orf5, CWC15, Cwf15, HSPC148, ORF5, Protein CWC15 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51503, 66070 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JHS9, Q9P013 | |
| CWC15 | |
| CWC15 Fusion Protein Ag18116 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction