missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CWC15 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44275CL1
This item is not returnable.
View return policy
Description
CWC15 Polyclonal antibody specifically detects CWC15 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| CWC15 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51503 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| C11orf5ORF5, CWC15 homolog, CWC15 homolog (S. cerevisiae), CWC15 spliceosome-associated protein homolog (S. cerevisiae), Cwf15, HSPC148, protein CWC15 homolog | |
| The epitope recognized by this antibody maps to a region between residue 75 to 125 of human Protein CWC15 homolog using the numbering given in entry Q9P013.2 (GeneID 51503). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction