missing translation for 'onlineSavingsMsg'
Learn More

CW-069, MedChemExpress

Product Code. 16436878
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16436878 10 mg 10mg
16429417 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16436878 Supplier Medchem Express Supplier No. HY15857
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CW-069 is an allosteric inhibitor of microtubule motor protein HSET with an IC50 of 75 μM.

CW-069 is an allosteric inhibitor of microtubule motor protein HSET with an IC50 of 75 μM.

Specifications

Chemical Name or Material CW-069
CAS 1594094-64-0
Color White
Purity Grade Notes Research
Molecular Formula C23H21IN2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 52 mg/mL (103.93 mM)
SMILES O=C(NC1=CC=C(I)C=C1C(O)=O)[C@@H](NCC2=CC=CC=C2)CC3=CC=CC=C3
Molecular Weight (g/mol) 500.33
Formula Weight 500.33
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.