Learn More
Invitrogen™ CUX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113847
Description
Immunogen sequence: SLGTEQSYPS PQQLPPPPGP EDPLSPSPGQ PLLGPSLGPD GTRTFSLSPF For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
CUX2 may be a transcription factor involved in neural specification. It binds to DNA in a sequence-specific manner.
Specifications
| CUX2 | |
| Polyclonal | |
| Unconjugated | |
| CUX2 | |
| 1700051K22Rik; CDP2; cut like homeobox 2; Cutl2; cut-like 2; cut-like homeobox 2; CUX2; Cux-2; ENSMUSG00000072641; homeobox protein cut-like 2; Homeobox protein cux-2; KIAA0293 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23316 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O14529 | |
| CUX2 | |
| Synthetic peptide directed towards the N terminal region of human CUX2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.