missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CUX1 Recombinant Rabbit Monoclonal Antibody (K01_2P09)

Product Code. 30829999
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829999 100 μL
30828896 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829999 Supplier Invitrogen™ Supplier No. MA564177
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: CAELQVRITE AVATATEQRE LIARLEQDLS IIQSIQRPDA EGAAEHRLEK IPEPIKEATA LFYGPAAPAS GALPEGQVDS LLSIISSQRE RFRARNQELE AENRLAQHTL QALQSELDSL RADNIKLFEK IKFLQSYPGR GSGSDDTELR YSSQYEERLD PFSSFSKRER QRKYLSLSPW DKATLSMGRL VLSNKMART Highest antigen sequence identity to the following orthologs: Mouse - 87%, Pig - 93%, Cynomolgus monkey - 99%.

The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUX1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2P09
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CUX1
Gene Accession No. P39880, P53565, P70403, Q13948
Gene Alias CASP; CCAAT displacement protein; CDP; CDP/Cut; CDP1; CDP2; Cdpl1; Clox; COY1; cut homolog; cut like homeobox 1; CUTL1; cut-like 1; cut-like homeobox 1; Cux; Cux/CDP; Cux/CDP homeoprotein; CUX1; Cux-1; golgi integral membrane protein 6; GOLIM6; Homeobox protein cut-like 1; homeobox protein cux-1; Kiaa4047; Nbla10317; p100; p110; p200; p75; Protein CASP; putative protein product of Nbla10317
Gene Symbols CUX1
Host Species Rabbit
Immunogen Recombinant protein of human Protein CASP (aa 421-619).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116639, 13047, 1523
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.