missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Cuvettes for Horiba ABX Pentra 400

Product Code. 31922710
Change view
Click to view available options
Quantity:
450/Cs.
Unit Size:
Pack of 450
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31922710 450/Cs. Pack of 450
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922710 Supplier Globe Scientific Supplier No. 5162
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed specifically for use with the Horiba Pentra 400 chemistry analyzers. Globe Scientific Cuvettes for Horiba ABX Pentra 400 provide a great alternative to OEM cuvettes. Cuvette segments are constructed from polymethylmethacrylate.

  • For use with the Horiba Pentra 400 chemistry analyzers
  • Great alternative to OEM cuvettes
  • Cuvette segments constructed from polymethylmethacrylate (PMMA)
  • Natural color

Specifications

Color Natural
Quantity 450/Cs.
Material PMMA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.