missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Cuvette for Cobas Mira, case of 300

Product Code. 31922632 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
300/Cs.
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31922632 300/Cs. Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922632 Supplier Globe Scientific Supplier No. 5121
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use on the Cobas Mira, Mira S and Mira Plus analyzers. Globe Scientific Cuvettes for Cobas Mira come individually wrapped and are constructed of polymethylmethacrylate. These 12-place Cuvette segments are designed for compatibility with these products. Case of 300.

  • For use on the Cobas Mira, Mira S and Mira Plus analyzers
  • 12-place cuvette segments
  • Individually wrapped
  • Polymethylmethacrylate (PMMA)
  • Natural color
  • Case of 300

Specifications

For Use With (Equipment) Cobas Mira, Mira S, and Mira Plus Analyzers
Color Natural
Quantity 300/Cs.
Material PMMA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.