missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUTL2 Antibody (CL14332) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26842PE
This item is not returnable.
View return policy
Description
CUTL2 Monoclonal antibody specifically detects CUTL2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| CUTL2 | |
| Monoclonal | |
| PE | |
| CDP2, CUTL2, cut-like 2 (Drosophila), cut-like homeobox 2, homeobox protein cut-like 2, Homeobox protein cux-2, KIAA0293 | |
| This antibody was developed using a synthetic peptide derived from O14529, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CL14332 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 23316 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction