missing translation for 'onlineSavingsMsg'
Learn More

Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eBioscience™

Cutaneous Lymphocyte Antigen (CLA) Monoclonal Antibody (HECA-452), eBioscience™

Brand:  Affymetrix eBioscience 14-9857-37

 View more versions of this product

Product Code. 17180802

  • £1376.00 / 2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cutaneous Lymphocyte Antigen (CLA)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
2 mg
Primary
4°C
Liquid
Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Western Blot
HECA-452
Unconjugated
CLA
Affinity Chromatography
RUO
Human
Antibody
IgM κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.