missing translation for 'onlineSavingsMsg'
Learn More

Cutamesine dihydrochloride, MedChemExpress

Product Code. 16491877
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16491877

Brand: Medchem Express HY13510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cutamesine dihydrochloride (SA4503 dihydrochloride; AGY94806 dihydrochloride) is a potent Sigma 1 receptor agonist with an IC50 of 17.4 nM in guinea pig brain membranes.

Cutamesine dihydrochloride (SA4503 dihydrochloride

Specifications

Chemical Name or Material Cutamesine dihydrochloride
CAS 165377-44-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H34Cl2N2O2
Quantity 50 mg
Synonym SA4503 dihydrochloride AGY94806 dihydrochloride
Solubility Information H2O : 100 mg/mL (226.54 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (67.96 mM; Need ultrasonic and warming)
SMILES COC1=CC=C(CCN2CCN(CCCC3=CC=CC=C3)CC2)C=C1OC.Cl.Cl
Molecular Weight (g/mol) 441.43
Formula Weight 441.43
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.