missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Custom TaqMan™ SNP Genotyping Assays

Product Code. 32226596 Shop All Applied Biosystems Products
Change view
Click to view available options
Sample Type:
Human or Non-human
Non-human
Quantity:
1000 Reactions, Made to Order
2400 Reactions, Made to Order
300 Reactions, Made to Order
L (2400 reactions), Made to Order
M (1000 reactions), Made to Order
S (300 reactions), Made to Order
Unit Size:
Each
Pack of 40
Pack of 80
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Type Quantity Unit Size
32226596 Human or Non-human 2400 Reactions, Made to Order Each
13435096 Non-human S (300 reactions), Made to Order Each
15531034 Non-human M (1000 reactions), Made to Order Pack of 40
15541034 Non-human L (2400 reactions), Made to Order Pack of 80
32226515 Human or Non-human 300 Reactions, Made to Order Each
32226521 Human or Non-human 1000 Reactions, Made to Order Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 32226596 Supplier Applied Biosystems™ Supplier No. A80739
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide a highly flexible technology for detection of polymorphisms within any genome.

Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5'-nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNA samples. Each assay enables genotyping of individuals for a single nucleotide polymorphism (SNP) and consists of two sequence-specific primers and two TaqMan minor groove binder (MGB) probes with non-fluorescent quenchers (NFQ). Assays contain one probe labeled with VIC or Cyanine 5 dye to detect the Allele 1 sequence; the second probe is labeled with FAM or ABY dye to detect the Allele 2 sequence.

Custom TaqMan SNP Genotyping Assays can be easily designed at no extra charge by submitting target sequences confidentially to our secure Custom Assay Design Tool.

Benefits:

  • Proven—gold-standard TaqMan chemistry and robust assay designs deliver accurate, reproducible, and reliable results
  • Easy—convenient single-tube format and simple workflow provide an easy path to trusted results; no optimization required
  • Flexible—obtain assay designs for any SNP in the human genome using our secure Custom Assay Design Tool, at no extra charge
  • Tested—all custom human assays (VIC/FAM SKUs only) are subjected to a functional test on 20 unique DNA samples. Assays ordered with non-human or Cyanine 5/ABY SKUs are not functionally tested.
  • Multiplexing ready—TaqMan assays are now available with with VIC/FAM or Cyanine 5/ABY formats to support simultaneous detection of two sets of SNPs (two pairs of alleles).

Approximate ship time

4–6 days in North America and 6–10 days in Europe (VIC/FAM); 4-10 days in North America, 6-14 days in Europe (Cyanine 5/ABY)

The free Custom Assay Design Tool can generate assays for the detection of SNPs, MNPs, and indels of up to six bases. These custom assays are designed, synthesized, formulated, optimized, and quality control tested.

TaqMan SNP Genotyping Assays require only three reaction components for PCR: purified genomic DNA (1–20 ng), the assay solution, and TaqMan Genotyping Master Mix (singleplex) or TaqPath ProAmp Master Mix (multiplex) (sold separately).

All assay designs are the product of our industry-leading bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355) or TaqPath ProAmp Master Mix (Cat. No. A30865)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 80X
Content And Storage •1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).
•Store at -15°C to -25°C.
For Use With (Application) Genotyping
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne, StepOnePlus, ViiA 7 System, QuantStudio Absolute Q Digital PCR System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler** If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals.
Key Functions Cyanine 5 - MGB probe detects Allele 1, ABY - MGB probe detects Allele 2
No. of Reactions 2400 Reactions, Made to Order
Product Line TaqMan
Product Type SNP Genotyping
Quantity 2400 Reactions, Made to Order
Shipping Condition Room Temperature
Format Tube
Sample Type Human or Non-human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.