Learn More
Ambion™ Custom Silencer™ siRNA
Custom Silencer™ siRNA
Brand:  Ambion™ AM16106
Code : NEW
Additional Details : Weight : 0.00450kg
 View more versions of this product
Product Code. 15551144
  • £235.00 / 40nmol
Estimated Shipment: 28-05-2024
to see stock.
Description
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Specifications
Room temperature
Custom siRNA
40 nmol
Custom siRNA, 40nmol, Liquid, Standard Purity, Water Soluble, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Standard