Learn More
Invitrogen™ Custom Silencer™ siRNA
Product Code. 15571064
Change view
Click to view available options
Description:
Custom siRNA HPLC, 250 nmol
Custom siRNA, 10 umol
Custom siRNA, 20 nmol
Custom siRNA, 250 nmol
Unit Size:
10µmol
20nmol
250nmol
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15571064 Custom siRNA HPLC, 250 nmol 250nmol
15531481 Custom siRNA HPLC, 250 nmol Each
15531144 Custom siRNA, 20 nmol 20nmol
15511154 Custom siRNA, 250 nmol 250nmol
15531154 Custom siRNA, 10 umol 10µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15571064 Supplier Invitrogen™ Supplier No. 4404015
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types.
Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store at or below -20°C. Do not store in a frost-free freezer. Dried oligonucleotides are shipped at ambient temperature.
Description Custom siRNA HPLC, 250 nmol
RNAi Type siRNA
Product Line Ambion™, Silencer™ Select
Product Type siRNA
Purity HPLC
Quantity 250 nmol
For Research Use Only. Not for use in diagnostic procedures.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.