missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom RNA, HPLC

Product Code. 10686983
Click to view available options
Product Type:
RNA HPLC
siRNA HPLC
Quantity:
20 nmol
20 nmol, 1 vial
5 nmol
50 nmol
500 nmol
80 nmol, 1 vial
Unit Size:
20nmol
50nmol
80nmol
Each
This item is not returnable. View return policy

Product Code. 10686983

Brand: Ambion™ 100001849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Custom RNA, HPLC

Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description RNA HPLC, 20nmol, Pre Strand, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA HPLC
Purity HPLC
Quantity 20 nmol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.