Learn More
Ambion™ Custom RNA
Product Code. 12037626
Change view
Click to view available options
Product Type:
RNA
RNA DSL In-vivo
RNA In-vivo
Quantity:
1 μmol
1.7 μmol
100 nmol
20 nmol
5 nmol
80 nmol
Unit Size:
1µmol
20nmol
80nmol
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
12037626 RNA 20 nmol 20nmol
10789534 RNA 80 nmol 80nmol
10206252 RNA 1 μmol 1µmol
10164322 RNA 1.7 μmol Each
10584414 RNA In-vivo 20 nmol Each
10647353 RNA In-vivo 5 nmol Each
10308323 RNA DSL In-vivo 100 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 12037626 Supplier Ambion™ Supplier No. 10620000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Research Use Only. All usage must comply with product instructions.
Custom RNA
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism. For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.
TRUSTED_SUSTAINABILITY
Specifications
Description Block-iT™ RNAi Designer, 20nmol, For Stealth RNAi™, Specific Gene Expression Knockdowns, Enhanced Target Specificity
Product Type RNA
Quantity 20 nmol