Learn More
Custom Primer
Product Code. 17886481
Change view
Click to view available options
Unit Size:
100nmol
Product Code. Unit Size
17886481 100nmol
1 options
Product Code. 17886481 Supplier Invitrogen Supplier No. 11732021
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.