missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Custom PreAmp Pools
Shop All Applied Biosystems Products
Click to view available options
Format:
96A
96B
Format 12
Format 16
Format 24
Format 32
Format 384
Format 48
Format 64
Unit Size:
1mL
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
Specifications
Specifications
| Type | Custom TaqMan™ miRNA Pre-Amp Pool |
| Format | Format 32 |
| Content And Storage | Room Temperature |
| No. of Samples | 1 to 8 |
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction