missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs
Shop All Applied Biosystems ProductsDescription
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
Specifications
Specifications
| Description | Custom 5' Labeled/Unlabeled Di-Repeat + Tail, 10000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications |
| Product Type | Custom 5' Labeled or Unlabeled Di-Repeat + Tail |
| Content And Storage | -20°C |
| Quantity | 10,000 pmol |
| For Use With (Application) | Fragment Analysis, Gene Expression Studies, Genotyping, PCR |
| Form | Delivered either in Solution or Dried down |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?