missing translation for 'onlineSavingsMsg'
Learn More

Custom 5' Fluorescent Labeled/Unlabeled Pairs

Product Code. 15507290
Change view
Click to view available options
300,000 pmol
80,000 pmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15507290 80,000 pmol
15537290 300,000 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15507290 Supplier Invitrogen Supplier No. 450059
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC®, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Oligonucleotides
Length (Metric) 1.524 m
Content And Storage Store in Freezer (-5°C to -30°C).
Purification Method Desalted
Label or Dye 6-FAM, HEX, NED, PET, TET, VIC
Primer 5' Fluorescent Labeled/Unlabeled
Quantity 80,000 picomoles⁄each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.