missing translation for 'onlineSavingsMsg'
Learn More

Cuprous Oxide, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15527934 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15527934 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15527934 Supplier Avantor J.T.Baker Supplier No. 187801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 1317-39-1
Physical Form Powder
Quantity 500 g
Molecular Formula Cu2O
Formula Weight 143.09
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 6.00kg
Chemical Name or Material Cuprous Oxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: copper(I) oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.